siRNA Details
Gene Symbol: C78197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030785.1 | - | 1 | 2249 | ||
AK037678.1 | - | 2 | 2000 | |||
BG079412.2 | - | 1 | 39 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C78197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030785.1 | - | 1 | 2249 | ||
AK037678.1 | - | 2 | 2000 | |||
BG079412.2 | - | 1 | 39 |