siRNA Details
| Gene Symbol: Gm31752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076902.1 | - | 2 | 80 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615671.1 | - | 2 | 77 | Chr. 5 - 18481907 - 18495258 [-] on Build GRCm38 | |

Gm31752
Genomic Map