siRNA Details
| Gene Symbol: Gm31752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076902.1 | - | 7 | 790 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV269595.2 | - | 6 | 609 | Chr. 5 - 18481710 - 18490019 [-] on Build GRCm38 | |

Gm31752
Genomic Map