siRNA Details
| Gene Symbol: AI504432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033498.1 | 2 | 3639 | |||
| GenBank mRNA | AK030783.1 | - | 1 | 1263 | ||
| AK046860.1 | - | 1 | 3113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI504432| Gene Symbol: AI504432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033498.1 | 2 | 3639 | |||
| GenBank mRNA | AK030783.1 | - | 1 | 1263 | ||
| AK046860.1 | - | 1 | 3113 | |||