siRNA Details
| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008235.1 | - | 1 | 1132 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB566242.1 | - | 1 | 457 | Chr. 10 - 81233855 - 81234716 [+] on Build GRCm38 | |

Zfr2
Genomic Map