siRNA Details
| Gene Symbol: 1110001D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003212.1 | - | 1 | 374 | ||
| AV006698.1 | - | 1 | 241 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703524.1 | - | 1 | 372 | Chr. 4 - 151932378 - 151932849 [-] on Build GRCm38 | |

1110001D16Rik
Genomic Map