siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045063.1 | 7 | 2110 | |||
| GenBank mRNA | AK030258.1 | - | 7 | 2111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930556M19Rik| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045063.1 | 7 | 2110 | |||
| GenBank mRNA | AK030258.1 | - | 7 | 2111 | ||