siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045063.1 | 2 | 225 | |||
| GenBank mRNA | AK030258.1 | - | 2 | 226 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB617275.1 | - | 2 | 226 | Chr. 15 - 10714836 - 10769215 [+] on Build GRCm38 | |

4930556M19Rik
Genomic Map