siRNA Details
| Gene Symbol: Gm30789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049712.1 | - | 1 | 2823 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm30789| Gene Symbol: Gm30789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049712.1 | - | 1 | 2823 | ||