siRNA Details
Gene Symbol: 4930470H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045764.1 | 2 | 1285 | |||
GenBank mRNA | AK015535.1 | - | 2 | 1287 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930470H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045764.1 | 2 | 1285 | |||
GenBank mRNA | AK015535.1 | - | 2 | 1287 |