siRNA Details
| Gene Symbol: 4933400L20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045730.1 | 5 | 664 | |||
| GenBank mRNA | AK016594.1 | - | 5 | 665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933400L20Rik| Gene Symbol: 4933400L20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045730.1 | 5 | 664 | |||
| GenBank mRNA | AK016594.1 | - | 5 | 665 | ||