siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019287.1 | - | 1 | 59 | ||
| AK028267.1 | - | 1 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp827| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019287.1 | - | 1 | 59 | ||
| AK028267.1 | - | 1 | 107 | |||