siRNA Details
| Gene Symbol: Slc35e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087552.1 | - | 1 | 660 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA619912.1 | - | 1 | 480 | Chr. 4 - 155601790 - 155602342 [+] on Build GRCm38 | |

Slc35e2
Genomic Map