siRNA Details
| Gene Symbol: BB155983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BB155983.1 | - | 1 | 239 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK137826.1 | - | 1100 | Chr. - - [-] on Build | ||

BB155983
Genomic Map