siRNA Details
Gene Symbol: Zfp963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK162670.1 | - | 4 | 1793 | ||
BC037669.1 | - | 2 | 1505 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK162670.1 | - | 4 | 1793 | ||
BC037669.1 | - | 2 | 1505 |