siRNA Details
| Gene Symbol: 4930572K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045371.1 | 2 | 82 | |||
| GenBank mRNA | AK016281.1 | - | 2 | 83 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715905.1 | - | 2 | 83 | Chr. 5 - 127170565 - 127184335 [+] on Build GRCm38 | |

4930572K03Rik
Genomic Map