siRNA Details
| Gene Symbol: 4930572K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045371.1 | 3 | 272 | |||
| GenBank mRNA | AK016281.1 | - | 3 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930572K03Rik| Gene Symbol: 4930572K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045371.1 | 3 | 272 | |||
| GenBank mRNA | AK016281.1 | - | 3 | 273 | ||