siRNA Details
| Gene Symbol: 4930572K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045371.1 | 5 | 793 | |||
| GenBank mRNA | AK016281.1 | - | 5 | 794 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464709.1 | - | 4 | 627 | Chr. 5 - 127170560 - 127184342 [+] on Build GRCm38 | |

4930572K03Rik
Genomic Map