siRNA Details
| Gene Symbol: Gm3258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | EL607674.1 | - | 1 | 625 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK139621.1 | - | 730 | Chr. - - [-] on Build | ||

Gm3258
Genomic Map