siRNA Details
| Gene Symbol: 4930551E15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019758.1 | - | 1 | 381 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715694.1 | - | 1 | 381 | Chr. 7 - 124186727 - 124187128 [+] on Build GRCm38 | |

4930551E15Rik
Genomic Map