siRNA Details
| Gene Symbol: 2900056B19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013703.1 | - | 1 | 594 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV153561.1 | - | 1 | 299 | Chr. 5 - 102725137 - 102725531 [-] on Build GRCm38 | |

2900056B19Rik
Genomic Map