siRNA Details
| Gene Symbol: 4930524B17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015880.1 | - | 5 | 617 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715434.1 | - | 5 | 617 | Chr. 6 - 19249065 - 19278843 [-] on Build GRCm38 | |

4930524B17Rik
Genomic Map