siRNA Details
| Gene Symbol: 4930548G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045811.1 | 2 | 241 | |||
| GenBank mRNA | AK016069.1 | - | 2 | 242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930548G14Rik| Gene Symbol: 4930548G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045811.1 | 2 | 241 | |||
| GenBank mRNA | AK016069.1 | - | 2 | 242 | ||