siRNA Details
| Gene Symbol: C430002E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040385.1 | 1, 2 | 120 | |||
| GenBank mRNA | AK021232.1 | - | 1, 2 | 122 | ||
| AK145605.1 | - | 1, 2 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C430002E04Rik| Gene Symbol: C430002E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040385.1 | 1, 2 | 120 | |||
| GenBank mRNA | AK021232.1 | - | 1, 2 | 122 | ||
| AK145605.1 | - | 1, 2 | 111 | |||