siRNA Details
| Gene Symbol: C430004N12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021233.1 | - | 1 | 306 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728938.1 | - | 1 | 303 | Chr. X - 152750974 - 152751588 [-] on Build GRCm38 | |

C430004N12Rik
Genomic Map