siRNA Details
Gene Symbol: C430004N12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021233.1 | - | 1 | 452 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728938.1 | - | 1 | 449 | Chr. X - 152750974 - 152751588 [-] on Build GRCm38 |
