siRNA Details
| Gene Symbol: C430005N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021234.1 | - | 1 | 456 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728941.1 | - | 1 | 454 | Chr. 19 - 17632881 - 17633490 [-] on Build GRCm38 | |

C430005N20Rik
Genomic Map