siRNA Details
| Gene Symbol: 2010106G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008322.1 | - | 1 | 84 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708250.1 | - | 1 | 81 | Chr. 17 - 29614465 - 29614769 [-] on Build GRCm38 | |

2010106G04Rik
Genomic Map