siRNA Details
| Gene Symbol: 1810012K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045473.1 | 3 | 510 | |||
| GenBank mRNA | AK007460.1 | - | 3 | 511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810012K16Rik| Gene Symbol: 1810012K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045473.1 | 3 | 510 | |||
| GenBank mRNA | AK007460.1 | - | 3 | 511 | ||