siRNA Details
| Gene Symbol: 1810012K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045473.1 | 1 | 155 | |||
| GenBank mRNA | AK007460.1 | - | 1 | 156 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707529.1 | - | 1 | 156 | Chr. 8 - 22398721 - 22399594 [+] on Build GRCm38 | |

1810012K16Rik
Genomic Map