siRNA Details
| Gene Symbol: Gm16499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039090.1 | - | 929 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CO803841.1 | - | 3 | 447 | Chr. 6 - 53858528 - 53867142 [+] on Build GRCm38 | |

Gm16499
Genomic Map