siRNA Details
| Gene Symbol: Mllt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085252.1 | - | 3 | 2853 | ||
| AK085575.1 | - | 1 | 1878 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mllt10| Gene Symbol: Mllt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085252.1 | - | 3 | 2853 | ||
| AK085575.1 | - | 1 | 1878 | |||