siRNA Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047449.1 | - | 2 | 1642 | ||
| AK049811.1 | - | 2 | 1586 | |||
| AK078269.1 | - | 1 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sybu| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047449.1 | - | 2 | 1642 | ||
| AK049811.1 | - | 2 | 1586 | |||
| AK078269.1 | - | 1 | 446 | |||