siRNA Details
| Gene Symbol: 9530006O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020548.1 | - | 1 | 249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530006O14Rik| Gene Symbol: 9530006O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020548.1 | - | 1 | 249 | ||