siRNA Details
| Gene Symbol: 1700025F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040578.1 | 2 | 219 | |||
| GenBank mRNA | AK006336.1 | - | 2 | 223 | ||
| AK015773.1 | - | 2 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700025F24Rik| Gene Symbol: 1700025F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040578.1 | 2 | 219 | |||
| GenBank mRNA | AK006336.1 | - | 2 | 223 | ||
| AK015773.1 | - | 2 | 198 | |||