siRNA Details
| Gene Symbol: Gm35215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054100.1 | - | 3 | 463 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB665859.1 | - | 3 | 460 | Chr. 13 - 98937797 - 98945915 [-] on Build GRCm38 | |

Gm35215
Genomic Map