siRNA Details
| Gene Symbol: Gm32014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110513.1 | 5 | 2933 | |||
| GenBank mRNA | AK054106.1 | - | 5 | 2933 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm32014| Gene Symbol: Gm32014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110513.1 | 5 | 2933 | |||
| GenBank mRNA | AK054106.1 | - | 5 | 2933 | ||