siRNA Details
| Gene Symbol: 2010203P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008426.1 | - | 2 | 340 | ||
| AK008432.1 | - | 2 | 340 | |||
| AV070026.1 | - | 1 | 133 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708335.1 | - | 2 | 340 | Chr. 13 - 56595537 - 56596642 [+] on Build GRCm38 | |

2010203P06Rik
Genomic Map