siRNA Details
| Gene Symbol: E030044B06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045343.1 | 4 | 378 | |||
| GenBank mRNA | AK087313.1 | - | 4 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030044B06Rik| Gene Symbol: E030044B06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045343.1 | 4 | 378 | |||
| GenBank mRNA | AK087313.1 | - | 4 | 379 | ||