siRNA Details
| Gene Symbol: E030044B06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045343.1 | 5 | 865 | |||
| GenBank mRNA | AK087313.1 | - | 5 | 866 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB535480.2 | - | 3 | 618 | Chr. 19 - 40876759 - 40882639 [+] on Build GRCm38 | |

E030044B06Rik
Genomic Map