siRNA Details
| Gene Symbol: 2810039B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012887.1 | - | 457 | |||
| AK084954.1 | - | 454 | ||||
| AK171817.1 | - | 101 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810039B14Rik| Gene Symbol: 2810039B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012887.1 | - | 457 | |||
| AK084954.1 | - | 454 | ||||
| AK171817.1 | - | 101 | ||||