siRNA Details
| Gene Symbol: 1700020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006151.1 | - | 2 | 253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706255.1 | - | 2 | 253 | Chr. 12 - 26572108 - 26602309 [-] on Build GRCm38 | |

1700020D12Rik
Genomic Map