siRNA Details
| Gene Symbol: 1700020G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006158.1 | - | 1 | 863 | ||
| BB806657.1 | - | 1 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700020G03Rik| Gene Symbol: 1700020G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006158.1 | - | 1 | 863 | ||
| BB806657.1 | - | 1 | 335 | |||