siRNA Details
| Gene Symbol: Tspan2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040588.1 | 3, 4 | 370 | |||
| GenBank mRNA | AK019118.1 | - | 3, 4 | 372 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710128.1 | - | 3, 4 | 372 | Chr. 3 - 102720231 - 102735419 [-] on Build GRCm38 | |

Tspan2os
Genomic Map