siRNA Details
| Gene Symbol: 2310002D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045490.1 | 3 | 397 | |||
| GenBank mRNA | AK009088.1 | - | 3 | 398 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708936.1 | - | 3 | 398 | Chr. 12 - 80507206 - 80517904 [+] on Build GRCm38 | |

2310002D06Rik
Genomic Map