siRNA Details
| Gene Symbol: 4930448I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040475.1 | 1 | 147 | |||
| GenBank mRNA | AK015419.1 | - | 1 | 148 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714967.1 | - | 1 | 148 | Chr. 1 - 41167560 - 41181252 [-] on Build GRCm38 | |

4930448I06Rik
Genomic Map