siRNA Details
| Gene Symbol: 4930405L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110478.1 | 1 | 54 | |||
| GenBank mRNA | AK019567.1 | - | 1 | 54 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714632.1 | - | 1 | 54 | Chr. 5 - 45926821 - 45932409 [-] on Build GRCm38 | |

4930405L22Rik
Genomic Map