siRNA Details
| Gene Symbol: 4930448C13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045960.1 | 1 | 24 | |||
| GenBank mRNA | AK015413.1 | - | 1 | 26 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714963.1 | - | 1 | 26 | Chr. 12 - 14996389 - 15050389 [-] on Build GRCm38 | |

4930448C13Rik
Genomic Map