siRNA Details
| Gene Symbol: 4930448D08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015414.1 | - | 2 | 443 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714964.1 | - | 2 | 443 | Chr. 18 - 69969677 - 69971817 [+] on Build GRCm38 | |

4930448D08Rik
Genomic Map