siRNA Details
| Gene Symbol: Gm36529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049017.1 | - | 4 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB650341.1 | - | 4 | 512 | Chr. 13 - 70840451 - 70858713 [+] on Build GRCm38 | |

Gm36529
Genomic Map